Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197216.2 | NP_001184145.1 | 8 | 1344 | ||
NM_001671.4 | NP_001662.1 | 9 | 1461 | |||
XM_011523861.2 | XP_011522163.1 | 9 | 1368 | |||
GenBank mRNA | AB070933.1 | - | 1240 | |||
AK308375.1 | - | 9 | 1265 | |||
BC032130.2 | - | 9 | 1138 | 5087444 | ||
BM818592.1 | - | 1 | 261 |