Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285.3 | NP_001276.2 | 2 | 401 | ||
GenBank mRNA | AF039400.1 | - | 3 | 623 | ||
AF127036.1 | - | 2 | 276 | |||
AK291999.1 | - | 2 | 317 | |||
AK314375.1 | - | 2 | 319 | |||
DA428844.1 | - | 2 | 401 |