Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001830.3 | NP_001821.2 | 4 | 600 | ||
GenBank mRNA | AB019432.1 | - | 3 | 445 | ||
AF170492.1 | - | 2 | 170 | |||
AK225287.1 | - | 4 | 561 | |||
AK289564.1 | - | 4 | 547 | |||
BC036068.1 | - | 3 | 220 | 5311366 | ||
BC130278.1 | - | 3 | 459 | 40146309 | ||
X77197.1 | - | 4 | 553 |