Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130698.1 | NP_001124170.1 | 6 | 1789 | ||
NM_003305.2 | NP_003296.1 | 5 | 1571 | |||
XM_011532217.2 | XP_011530519.1 | 6 | 2095 | |||
XM_011532218.2 | XP_011530520.1 | 6 | 2095 | |||
XM_017008578.1 | XP_016864067.1 | 6 | 2095 | |||
XM_017008579.1 | XP_016864068.1 | 6 | 2095 | |||
GenBank mRNA | AB255423.1 | - | 4 | 1331 | ||
AB255424.1 | - | 5 | 1496 | |||
AY865574.1 | - | 6 | 1789 | |||
BC093682.1 | - | 5 | 1571 | 7939527 | ||
BC093684.1 | - | 5 | 1571 | 7939529 | ||
Y13758.1 | - | 5 | 1920 |