Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014379.3 | NP_055194.1 | 4 | 3159 | ||
GenBank mRNA | AF167082.2 | - | 3 | 2467 | ||
AK308123.1 | - | 4 | 3152 |