Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101669.1 | NP_001095139.1 | 13, 14 | 1464 | ||
NM_003866.2 | NP_003857.2 | 14, 15 | 1563 | |||
XM_005263323.3 | XP_005263380.1 | 6, 7 | 672 | |||
XM_005263325.3 | XP_005263382.1 | 6, 7 | 645 | |||
XM_011532391.2 | XP_011530693.1 | 13, 14 | 1548 | |||
XM_011532392.2 | XP_011530694.1 | 13, 14 | 1314 | |||
XM_017008794.1 | XP_016864283.1 | 13, 14 | 1548 | |||
XM_017008795.1 | XP_016864284.1 | 13, 14 | 1548 | |||
XM_017008796.1 | XP_016864285.1 | 13, 14 | 1548 | |||
XM_017008797.1 | XP_016864286.1 | 6, 7 | 668 | |||
XM_017008798.1 | XP_016864287.1 | 6, 7 | 645 | |||
GenBank mRNA | AK298118.1 | - | 6, 7 | 613 | ||
AK300891.1 | - | 6, 7 | 678 | |||
AK302936.1 | - | 12, 13 | 1230 | |||
BC005274.1 | - | 13, 14 | 1239 | |||
BC072447.1 | - | 12, 13 | 1371 | |||
BC110918.1 | - | 12, 13 | 1321 | 6160633 | ||
BC133005.1 | - | 10, 11 | 1019 | 40147027 | ||
BC144255.1 | - | 10, 11 | 1019 | |||
BX649090.1 | - | 14, 15 | 1404 | |||
U96922.1 | - | 10, 11 | 1089 |