Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145443.2 | NP_001138915.1 | 13, 14 | 1409 | ||
NM_001282630.2 | NP_001269559.1 | 13, 14 | 1522 | |||
NM_001314063.1 | NP_001300992.1 | 13, 14 | 1733 | |||
NM_001323016.1 | NP_001309945.1 | 13, 14 | 1409 | |||
NM_001323017.1 | NP_001309946.1 | 13, 14 | 1783 | |||
NM_004566.3 | NP_004557.1 | 13, 14 | 1733 | |||
XM_017016325.1 | XP_016871814.1 | 13, 14 | 1733 | |||
XM_017016326.1 | XP_016871815.1 | 12, 13 | 1676 | |||
XM_017016327.1 | XP_016871816.1 | 13, 14 | 1485 | |||
XM_017016328.1 | XP_016871817.1 | 12, 13 | 1628 | |||
XM_017016329.1 | XP_016871818.1 | 12, 13 | 1676 | |||
GenBank mRNA | AF056320.1 | - | 14, 15 | 1393 | ||
AF109735.1 | - | 13, 14 | 1463 | |||
AJ272439.1 | - | 1, 2 | 56 | |||
AJ295747.1 | - | 1, 2 | 56 | |||
AK291263.1 | - | 13, 14 | 1678 | |||
AK303039.1 | - | 13, 14 | 1342 | |||
AK304450.1 | - | 13, 14 | 1454 | |||
BC037258.1 | - | 13, 14 | 1334 | |||
BC040482.1 | - | 13, 14 | 1678 | 5271907 | ||
D49817.1 | - | 13, 14 | 1367 | |||
L77662.1 | - | 13, 14 | 1687 |