Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167576.1 | NP_001161048.1 | 5 | 1698 | ||
NM_001167577.1 | NP_001161049.1 | 6 | 1863 | |||
NM_020389.2 | NP_065122.1 | 7 | 2046 | |||
GenBank mRNA | AJ272034.1 | - | 6 | 1824 | ||
AJ421783.1 | - | 6 | 1824 | |||
AJ549088.1 | - | 5 | 1641 | |||
AJ549089.1 | - | 5 | 1659 | |||
AJ549090.1 | - | 3 | 1259 | |||
AY167926.1 | - | 5 | 1656 | |||
AY167927.1 | - | 4 | 1476 | |||
BC128185.1 | - | 6 | 1824 | 40113370 | ||
BC128186.1 | - | 6 | 1840 |