Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004685.3 | NP_004676.3 | 14 | 3224 | ||
XM_011535306.1 | XP_011533608.1 | 15 | 3284 | |||
XM_011535307.1 | XP_011533609.1 | 14 | 3157 | |||
XM_017020845.1 | XP_016876334.1 | 13 | 3043 | |||
XM_017020846.1 | XP_016876335.1 | 11 | 2489 | |||
GenBank mRNA | AF072928.1 | - | 10 | 2438 | ||
AK025507.1 | - | 4 | 1613 | |||
AK094082.1 | - | 1 | 1592 | |||
AL832017.2 | - | 14 | 3147 | |||
BC031834.1 | - | 1 | 1187 | |||
BC035436.1 | - | 1 | 1187 | |||
BC040012.1 | - | 14 | 3096 | 5495564 | ||
U47635.1 | - | 3 | 1571 |