Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040142.1 | NP_001035232.1 | 27 | 6016 | ||
NM_001040143.1 | NP_001035233.1 | 26 | 5777 | |||
NM_021007.2 | NP_066287.2 | 27 | 6082 | |||
XM_005246753.3 | XP_005246810.1 | 27 | 6032 | |||
XM_005246755.4 | XP_005246812.1 | 22 | 5117 | |||
XM_017004655.1 | XP_016860144.1 | 27 | 5931 | |||
XM_017004656.1 | XP_016860145.1 | 27 | 5987 | |||
XM_017004657.1 | XP_016860146.1 | 27 | 5884 | |||
XM_017004658.1 | XP_016860147.1 | 27 | 6196 | |||
XM_017004659.1 | XP_016860148.1 | 15 | 5114 | |||
GenBank mRNA | AB208888.1 | - | 25 | 9328 | ||
M91804.1 | - | 1 | 603 | |||
M94055.1 | - | 27 | 5831 | |||
X65361.1 | - | 1 | 603 |