Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195001.1 | NP_001181930.1 | 30 | 5523 | ||
NM_005704.4 | NP_005695.3 | 31 | 5562 | |||
NM_133177.3 | NP_573438.3 | 31 | 5544 | |||
NM_133178.3 | NP_573439.2 | 30 | 5532 | |||
XM_006710269.2 | XP_006710332.1 | 31 | 5525 | |||
XM_016999992.1 | XP_016855481.1 | 32 | 5555 | |||
XM_016999993.1 | XP_016855482.1 | 32 | 5546 | |||
XM_016999994.1 | XP_016855483.1 | 31 | 5528 | |||
XM_016999995.1 | XP_016855484.1 | 27 | 4721 | |||
GenBank mRNA | AB208855.1 | - | 26 | 4826 | ||
AK094849.1 | - | 6 | 1659 | |||
BC146655.1 | - | 30 | 5490 | 40147934 | ||
U73727.1 | - | 30 | 5523 | |||
X97198.1 | - | 30 | 5513 |