Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006244.3 | NP_006235.1 | 12, 13 | 1860 | ||
XM_011545132.2 | XP_011543434.1 | 13, 14 | 1293 | |||
XM_011545133.1 | XP_011543435.1 | 14, 15 | 1413 | |||
XM_011545134.2 | XP_011543436.1 | 12, 13 | 2699 | |||
GenBank mRNA | BC045619.1 | - | 12, 13 | 1854 | 5294756 | |
L42374.1 | - | 12, 13 | 1563 |