Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256613.1 | NP_001243542.1 | 5 | 916 | ||
NM_001256614.1 | NP_001243543.1 | 3 | 722 | |||
NM_182589.2 | NP_872395.2 | 4 | 689 | |||
NM_198313.2 | NP_938055.1 | 4 | 871 | |||
NM_198314.2 | NP_938056.1 | 3 | 644 | |||
GenBank mRNA | AY159813.2 | - | 4 | 689 | ||
AY349352.1 | - | 4 | 405 | |||
AY349353.1 | - | 3 | 450 | |||
BC101182.1 | - | 4 | 636 | 40017961 | ||
BC101183.2 | - | 4 | 636 | 40017962 | ||
BC101185.2 | - | 5 | 468 | 40017964 | ||
DQ644022.1 | - | 5 | 916 | |||
EU165354.1 | - | 3 | 528 |