Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206994.1 | NP_001193923.1 | 10 | 3596 | ||
NM_001206995.1 | NP_001193924.1 | 10 | 3700 | |||
NM_001206996.1 | NP_001193925.1 | 10 | 3750 | |||
NM_020416.3 | NP_065149.2 | 9 | 3888 | |||
NM_181876.2 | NP_870991.1 | 9 | 3542 | |||
XM_005247979.4 | XP_005248036.1 | 5 | 3733 | |||
XM_011513495.1 | XP_011511797.1 | 9 | 3637 | |||
XM_011513496.1 | XP_011511798.1 | 9 | 3652 | |||
GenBank mRNA | AK127386.1 | - | 1 | 1195 | ||
BC021735.2 | - | 1 | 614 | |||
BC045682.1 | - | 1 | 1596 |