Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142353.2 | NP_001135825.1 | 14 | 2136 | ||
NM_001142354.2 | NP_001135826.1 | 13 | 2106 | |||
NM_021132.3 | NP_066955.1 | 14 | 2133 | |||
XM_005269944.1 | XP_005270001.1 | 13 | 2103 | |||
XM_017016379.1 | XP_016871868.1 | 13 | 2038 | |||
GenBank mRNA | AJ488506.1 | - | 13 | 2043 | ||
AK300701.1 | - | 6 | 929 | |||
BC028049.1 | - | 14 | 2124 | 4538822 | ||
BC035464.1 | - | 14 | 2091 | |||
M29551.1 | - | 14 | 2114 |