Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199797.1 | NP_001186726.1 | 10 | 2142 | ||
NM_002832.3 | NP_002823.3 | 10 | 2988 | |||
NM_080588.2 | NP_542155.1 | 9 | 2481 | |||
NR_037663.1 | 10 | 2935 | ||||
NR_037664.1 | 9 | 2804 | ||||
XM_011509819.1 | XP_011508121.1 | 11 | 2461 | |||
XM_011509820.2 | XP_011508122.1 | 11 | 2165 | |||
XM_011509821.1 | XP_011508123.1 | 11 | 2093 | |||
XM_011509822.2 | XP_011508124.1 | 11 | 1964 | |||
XM_011509824.1 | XP_011508126.1 | 10 | 1969 | |||
XM_017001935.1 | XP_016857424.1 | 11 | 2458 | |||
XM_017001936.1 | XP_016857425.1 | 10 | 2320 | |||
XM_017001937.1 | XP_016857426.1 | 10 | 2138 | |||
GenBank mRNA | AK127214.1 | - | 10 | 2981 | ||
BC001746.1 | - | 9 | 2481 | 3543111 | ||
D11327.1 | - | 11 | 1910 | |||
M64322.1 | - | 10 | 1986 |