Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130701.1 | NP_001124173.1 | 12 | 2264 | ||
NM_145251.3 | NP_660294.1 | 11 | 2512 | |||
XM_006720242.3 | XP_006720305.1 | 9 | 2132 | |||
XM_011537108.2 | XP_011535410.1 | 11 | 2179 | |||
XM_011537109.2 | XP_011535411.1 | 8 | 2073 | |||
GenBank mRNA | AF085865.1 | - | 1 | 341 | ||
AL832591.1 | - | 12 | 2251 | |||
AL832697.1 | - | 2 | 1654 | |||
BC020265.2 | - | 11 | 2465 | 3922829 | ||
BC146995.1 | - | 11 | 2403 | 9056923 | ||
BC146998.1 | - | 11 | 2404 | 9056926 | ||
BX647636.1 | - | 11 | 2474 |