Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253829.1 | NP_001240758.1 | 9 | 4395 | ||
NM_001253830.1 | NP_001240759.1 | 10 | 4441 | |||
NM_152422.4 | NP_689635.3 | 9 | 4389 | |||
NM_177995.2 | NP_818931.1 | 10 | 4471 | |||
XM_005251711.4 | XP_005251768.1 | 10 | 6413 | |||
XM_011518240.2 | XP_011516542.1 | 10 | 4221 | |||
XM_011518241.2 | XP_011516543.1 | 10 | 4175 | |||
XM_017014280.1 | XP_016869769.1 | 11 | 4280 | |||
XM_017014281.1 | XP_016869770.1 | 10 | 4199 | |||
XM_017014282.1 | XP_016869771.1 | 7 | 4302 | |||
GenBank mRNA | AK094773.1 | - | 4 | 2444 | ||
BC067120.1 | - | 10 | 4456 | 5539182 |