Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001783.3 | NP_001774.1 | 5 | 1231 | ||
NM_021601.3 | NP_067612.1 | 5 | 1117 | |||
GenBank mRNA | AF283770.3 | - | 5 | 1074 | ||
AK223371.1 | - | 5 | 1229 | |||
BC018964.2 | - | 2 | 253 | |||
M80462.1 | - | 5 | 1082 | |||
M86921.1 | - | 5 | 1078 | |||
S46706.1 | - | 5 | 1052 | |||
S75217.1 | - | 5 | 1042 |