Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146068.1 | NP_001139540.1 | 6, 7 | 820 | ||
NM_001748.4 | NP_001739.2 | 6, 7 | 1042 | |||
XM_011544286.2 | XP_011542588.1 | 6, 7 | 1219 | |||
GenBank mRNA | AB209855.1 | - | 6, 7 | 907 | ||
AF261089.1 | - | 6, 7 | 945 | |||
AK297800.1 | - | 6, 7 | 820 | |||
AK303952.1 | - | 3 | 416 | |||
AK315179.1 | - | 6, 7 | 944 | |||
AK316211.1 | - | 6, 7 | 863 | |||
BC007686.2 | - | 6, 7 | 1042 | |||
BC011828.2 | - | 6, 7 | 1042 | |||
BC021303.2 | - | 6, 7 | 936 | 5090865 | ||
DA448245.1 | - | 3, 4 | 401 | |||
M23254.1 | - | 6, 7 | 948 |