Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002787.4 | NP_002778.1 | 8 | 737 | ||
GenBank mRNA | AK290654.1 | - | 8 | 712 | ||
AU120930.1 | - | 8 | 713 | |||
BC002900.2 | - | 7 | 663 | |||
BC047697.1 | - | 8 | 691 | 5786518 | ||
CR450317.1 | - | 8 | 689 | |||
D00760.1 | - | 8 | 689 |