Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001805.3 | NP_001796.2 | 2 | 1113 | ||
GenBank mRNA | BC035797.1 | - | 2 | 696 | 5735297 | |
U48866.1 | - | 2 | 532 |