Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244606.1 | NP_001231535.1 | 7 | 1154 | ||
NM_032549.3 | NP_115938.1 | 6 | 1468 | |||
XM_005250630.3 | XP_005250687.1 | 8 | 1236 | |||
XM_011516605.2 | XP_011514907.1 | 8 | 1252 | |||
XM_011516609.2 | XP_011514911.1 | 6 | 1411 | |||
XM_011516613.2 | XP_011514915.1 | 5 | 1327 | |||
XM_017012705.1 | XP_016868194.1 | 7 | 1206 | |||
GenBank mRNA | AF359563.1 | - | 6 | 1468 |