Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 4 | 570 | ||
NM_020351.3 | NP_065084.2 | 3 | 508 | |||
GenBank mRNA | AY550974.1 | - | 2 | 368 | ||
BC013581.1 | - | 4 | 548 | 3875911 | ||
BT009917.1 | - | 1 | 311 | |||
CA393523.1 | - | 3 | 494 | |||
CN484310.1 | - | 3 | 494 |