Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112741.1 | NP_001106212.1 | 2 | 1458 | ||
NM_004976.4 | NP_004967.1 | 2 | 1458 | |||
XM_011520080.2 | XP_011518382.1 | 2 | 2596 | |||
GenBank mRNA | BC107129.2 | - | 2 | 1475 | 40024733 | |
L00621.1 | - | 2 | 1471 | |||
M96747.1 | - | 1 | 674 | |||
S56770.1 | - | 2 | 1471 |