Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017424.2 | NP_001017424.1 | 2 | 605 | ||
NM_001017425.2 | NP_001017425.2 | 2 | 406 | |||
NM_014217.3 | NP_055032.1 | 2 | 282 | |||
XM_011509522.2 | XP_011507824.1 | 2 | 1808 | |||
XM_011509523.1 | XP_011507825.1 | 2 | 210 | |||
XM_011509524.2 | XP_011507826.1 | 2 | 257 | |||
XM_017001248.1 | XP_016856737.1 | 3 | 993 | |||
XM_017001249.1 | XP_016856738.1 | 3 | 944 | |||
GenBank mRNA | AF004711.1 | - | 2 | 390 | ||
AF129399.1 | - | 1 | 192 | |||
AF171068.1 | - | 2 | 206 | |||
AK123409.1 | - | 2 | 294 | |||
AK291483.1 | - | 2 | 386 | |||
AK294780.1 | - | 2 | 388 | |||
AK315249.1 | - | 3 | 372 | |||
AU280191.1 | - | 2 | 348 | |||
AY552980.2 | - | 2 | 292 | |||
AY552981.1 | - | 2 | 297 | |||
BC069462.1 | - | 2 | 206 | 7262344 | ||
BC101693.1 | - | 2 | 251 | 8069199 | ||
BC101695.1 | - | 2 | 251 | 8069201 | ||
BC143586.1 | - | 2 | 252 | 9052097 | ||
CD625291.1 | - | 2 | 602 | |||
CD625297.1 | - | 2 | 599 | |||
CD625301.1 | - | 2 | 220 | |||
DR001358.1 | - | 2 | 416 | |||
EF165334.1 | - | 2 | 339 | |||
EF165335.1 | - | 2 | 304 | |||
KF182338.1 | - | 2 | 241 | |||
KF182339.1 | - | 2 | 215 |