Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282232.1 | NP_001269161.1 | 3 | 412 | ||
NM_001282233.1 | NP_001269162.1 | 2 | 269 | |||
NM_001282234.1 | NP_001269163.1 | 3 | 289 | |||
NM_002791.2 | NP_002782.1 | 3 | 364 | |||
NR_104110.1 | 3 | 364 | ||||
XM_017021468.1 | XP_016876957.1 | 3 | 1700 | |||
GenBank mRNA | AK298920.1 | - | 2 | 237 | ||
AK302008.1 | - | 3 | 214 | |||
AK313011.1 | - | 3 | 290 | |||
AK316223.1 | - | 3 | 278 | |||
BC002979.1 | - | 3 | 255 | 3542966 | ||
BC017882.1 | - | 3 | 277 | 4278372 | ||
BC022354.1 | - | 3 | 234 | 4292526 | ||
BC023659.1 | - | 3 | 282 | 4869452 | ||
BC070137.1 | - | 3 | 286 | 6458246 | ||
BG701535.1 | - | 3 | 346 | |||
BU198521.1 | - | 3 | 238 | |||
BU657545.1 | - | 3 | 364 | |||
CR456944.1 | - | 3 | 204 | |||
HY028439.1 | - | 3 | 247 | |||
X59417.1 | - | 3 | 266 | |||
X61972.1 | - | 2 | 129 |