Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002793.3 | NP_002784.1 | 3 | 357 | ||
GenBank mRNA | AB209078.1 | - | 3 | 287 | ||
AB451312.1 | - | 3 | 270 | |||
AB451442.1 | - | 3 | 270 | |||
AK023290.1 | - | 2 | 1485 | |||
AK058050.1 | - | 3 | 327 | |||
AK223193.1 | - | 3 | 308 | |||
BC000508.2 | - | 3 | 308 | 2822268 | ||
BC020807.1 | - | 3 | 308 | 4281426 | ||
BP369175.1 | - | 3 | 369 | |||
BQ685374.1 | - | 2 | 87 | |||
BT019720.1 | - | 3 | 270 | |||
D00761.1 | - | 3 | 282 |