Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007058.3 | NP_008989.2 | 3, 4 | 376 | ||
XM_006714984.1 | XP_006715047.1 | 3, 4 | 379 | |||
XM_006714985.1 | XP_006715048.1 | 3, 4 | 379 | |||
XM_006714987.1 | XP_006715050.1 | 3, 4 | 379 | |||
XM_011514274.1 | XP_011512576.1 | 3, 4 | 460 | |||
XM_011514275.1 | XP_011512577.1 | 4, 5 | 502 | |||
GenBank mRNA | AJ242832.1 | - | 2, 3 | 330 | ||
AK314054.1 | - | 3, 4 | 356 | |||
BC033733.1 | - | 3, 4 | 347 | 5170266 | ||
BG721593.1 | - | 3, 4 | 381 | |||
DB052011.1 | - | 4, 5 | 466 |