Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311182.1 | NP_001298111.1 | 7 | 965 | ||
NM_022046.5 | NP_071329.2 | 7 | 965 | |||
GenBank mRNA | AF283670.1 | - | 6 | 824 | ||
BC074904.2 | - | 6 | 781 | 30915253 | ||
BC074905.2 | - | 6 | 781 | 30915482 | ||
BC114614.1 | - | 7 | 965 | 40031928 |