Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139159.4 | NP_631898.3 | 8 | 1098 | ||
XM_005259673.2 | XP_005259730.1 | 9 | 1207 | |||
XM_011528405.1 | XP_011526707.1 | 10 | 1259 | |||
XM_011528406.1 | XP_011526708.1 | 11 | 1369 | |||
XM_011528408.1 | XP_011526710.1 | 8 | 1044 | |||
XM_011528410.1 | XP_011526712.1 | 9 | 1259 | |||
XM_017027453.1 | XP_016882942.1 | 10 | 1849 | |||
XM_017027454.1 | XP_016882943.1 | 11 | 1959 | |||
GenBank mRNA | AF452102.2 | - | 8 | 1118 | ||
AF542510.2 | - | 8 | 1077 | |||
AK122645.1 | - | 2 | 148 | |||
AK122654.1 | - | 8 | 1101 | |||
AK131100.1 | - | 5 | 667 | |||
AK131499.1 | - | 7 | 611 | |||
AL834376.1 | - | 2 | 129 | |||
AY172660.1 | - | 5 | 763 | |||
AY374518.1 | - | 5 | 753 | |||
BC037948.2 | - | 8 | 1035 | 4577856 | ||
DQ417928.1 | - | 6 | 840 |