Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153612.3 | NP_705840.2 | 2 | 676 | ||
XM_006715379.2 | XP_006715442.1 | 5 | 1164 | |||
XM_011535588.2 | XP_011533890.1 | 3 | 1880 | |||
XM_017010470.1 | XP_016865959.1 | 5 | 1738 | |||
XM_017010471.1 | XP_016865960.1 | 5 | 2697 | |||
XM_017010472.1 | XP_016865961.1 | 4 | 2334 | |||
XM_017010473.1 | XP_016865962.1 | 5 | 1734 | |||
XM_017010474.1 | XP_016865963.1 | 5 | 1172 | |||
GenBank mRNA | AF503292.1 | - | 2 | 644 | ||
AK091074.1 | - | 1 | 193 | |||
AK126930.1 | - | 2 | 1336 | |||
AK289907.1 | - | 5 | 1163 | |||
BC093911.1 | - | 2 | 677 | 7939756 | ||
BC093913.1 | - | 2 | 677 | 7939758 |