Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001918.3 | NP_001909.3 | 3 | 230 | ||
XM_005270545.3 | XP_005270602.1 | 4 | 428 | |||
XM_017000468.1 | XP_016855957.1 | 5 | 457 | |||
XM_017000469.1 | XP_016855958.1 | 2 | 67 | |||
GenBank mRNA | AK303941.1 | - | 4 | 401 | ||
AK313191.1 | - | 3 | 212 | |||
BC016675.1 | - | 3 | 197 | 3924492 | ||
BC030002.1 | - | 3 | 197 | |||
BT007372.1 | - | 3 | 197 | |||
DA274670.1 | - | 3 | 230 | |||
J03208.1 | - | 3 | 195 | |||
M19301.1 | - | 3 | 195 | |||
M27093.1 | - | 3 | 195 | |||
X66785.1 | - | 3 | 211 |