Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001794.4 | NP_001785.2 | 2 | 192 | ||
GenBank mRNA | AK315192.1 | - | 2 | 146 | ||
BC101651.1 | - | 2 | 192 | 8069157 | ||
BC112150.1 | - | 2 | 192 | 8327618 | ||
L34059.1 | - | 2 | 118 |