Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000346.3 | NP_000337.1 | 3 | 2074 | ||
GenBank mRNA | AK295455.1 | - | 4 | 1880 | ||
BC007951.2 | - | 3 | 1931 | 4299305 | ||
BC056420.1 | - | 3 | 2084 | 6200521 | ||
Z46629.1 | - | 3 | 2061 |