Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000408.4 | NP_000399.3 | 12, 13 | 1750 | ||
NM_001083112.2 | NP_001076581.2 | 12, 13 | 1976 | |||
XM_005246469.2 | XP_005246526.1 | 12, 13 | 1689 | |||
XM_011510977.2 | XP_011509279.1 | 12, 13 | 1744 | |||
XM_011510978.2 | XP_011509280.1 | 13, 14 | 1794 | |||
XM_011510980.2 | XP_011509282.1 | 7, 8 | 975 | |||
XM_017003830.1 | XP_016859319.1 | 12, 13 | 1830 | |||
XM_017003831.1 | XP_016859320.1 | 11, 12 | 1639 | |||
GenBank mRNA | AB209399.1 | - | 12, 13 | 1745 | ||
AF311325.1 | - | 13, 14 | 1786 | |||
AK290057.1 | - | 12, 13 | 1911 | |||
AK291065.1 | - | 12, 13 | 1989 | |||
AK292817.1 | - | 12, 13 | 1749 | |||
AK299645.1 | - | 8, 9 | 1091 | |||
U12424.1 | - | 12, 13 | 1739 | |||
U36310.1 | - | 12, 13 | 1719 |