Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000408.4 | NP_000399.3 | 13 | 1878 | ||
NM_001083112.2 | NP_001076581.2 | 13 | 2104 | |||
XM_005246469.2 | XP_005246526.1 | 13 | 1817 | |||
XM_011510977.2 | XP_011509279.1 | 13 | 1872 | |||
XM_011510978.2 | XP_011509280.1 | 14 | 1922 | |||
XM_011510980.2 | XP_011509282.1 | 8 | 1103 | |||
XM_017003830.1 | XP_016859319.1 | 13 | 1958 | |||
XM_017003831.1 | XP_016859320.1 | 12 | 1767 | |||
GenBank mRNA | AB209399.1 | - | 13 | 1873 | ||
AF311325.1 | - | 14 | 1914 | |||
AK093198.1 | - | 14 | 1913 | |||
AK290057.1 | - | 13 | 2039 | |||
AK291065.1 | - | 13 | 2117 | |||
AK292817.1 | - | 13 | 1877 | |||
AK299645.1 | - | 9 | 1219 | |||
U12424.1 | - | 13 | 1867 | |||
U36310.1 | - | 13 | 1847 |