Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042426.1 | NP_001035891.1 | 2 | 337 | ||
NM_001809.3 | NP_001800.1 | 2 | 338 | |||
GenBank mRNA | BC000881.1 | - | 2 | 284 | 3461992 | |
BC002703.2 | - | 2 | 209 | 3626578 | ||
BT007246.1 | - | 2 | 155 | |||
U14518.1 | - | 2 | 298 |