Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000616.4 | NP_000607.1 | 3, 4 | 480 | ||
GenBank mRNA | AK308344.1 | - | 3, 4 | 323 | ||
AK312828.1 | - | 3, 4 | 417 | |||
BC025782.1 | - | 3, 4 | 374 | 5226427 | ||
BT019791.1 | - | 2, 3 | 222 | |||
BT019811.1 | - | 2, 3 | 222 | |||
DA384217.1 | - | 3, 4 | 481 | |||
DA463597.1 | - | 3, 4 | 467 | |||
M12807.1 | - | 2, 3 | 297 |