Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000817.2 | NP_000808.2 | 5 | 760 | ||
NM_013445.3 | NP_038473.2 | 5 | 764 | |||
XM_005246444.2 | XP_005246501.1 | 5 | 758 | |||
XM_011510922.1 | XP_011509224.1 | 5 | 614 | |||
XM_017003756.1 | XP_016859245.1 | 5 | 563 | |||
XM_017003757.1 | XP_016859246.1 | 5 | 758 | |||
XM_017003758.1 | XP_016859247.1 | 5 | 758 | |||
GenBank mRNA | A28074.1 | - | 4 | 338 | ||
AF178853.2 | - | 4 | 338 | |||
AK292582.1 | - | 5 | 563 | |||
BC002815.2 | - | 5 | 542 | |||
BC026349.1 | - | 5 | 564 | 4794214 | ||
BC036552.1 | - | 4 | 1167 | 5264831 | ||
BC037780.2 | - | 5 | 760 | 5273792 | ||
KU178086.1 | - | 4 | 338 | |||
L16888.1 | - | 5 | 521 | |||
M81883.1 | - | 5 | 888 | |||
S61897.1 | - | 4 | 347 | |||
Z22750.1 | - | 5 | 521 |