Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286055.1 | NP_001272984.1 | 2 | 451 | ||
NM_001286056.1 | NP_001272985.1 | 2 | 241 | |||
NM_001839.4 | NP_001830.1 | 2 | 451 | |||
XM_017000245.1 | XP_016855734.1 | 4 | 2863 | |||
GenBank mRNA | AK294141.1 | - | 2 | 202 | ||
AK298145.1 | - | 2 | 371 | |||
AK312370.1 | - | 2 | 338 | |||
AK316398.1 | - | 2 | 217 | |||
BC025372.2 | - | 2 | 335 | 3505668 | ||
BU149686.1 | - | 2 | 387 | |||
CR541848.1 | - | 2 | 122 | |||
DC422152.1 | - | 2 | 407 | |||
DR157009.1 | - | 2 | 374 | |||
S80562.1 | - | 2 | 205 |