Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 25 | 3532 | ||
GenBank mRNA | AB208885.1 | - | 22 | 2310 | ||
AJ510248.1 | - | 2 | 197 | |||
AJ510249.1 | - | 2 | 197 | |||
BC080190.1 | - | 8 | 877 | |||
L12002.1 | - | 25 | 3197 | |||
X16983.1 | - | 25 | 2811 |