Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 4, 5 | 589 | ||
NM_020351.3 | NP_065084.2 | 3, 4 | 527 | |||
GenBank mRNA | BC013581.1 | - | 4, 5 | 567 | 3875911 | |
BT009917.1 | - | 1, 2 | 330 | |||
CA393523.1 | - | 513 | ||||
CN484310.1 | - | 3, 4 | 513 | |||
X57527.1 | - | 1, 2 | 330 |