Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001186.3 | NP_001177.1 | 5 | 4624 | ||
NM_206866.2 | NP_996749.1 | 5 | 4648 | |||
GenBank mRNA | AB002803.1 | - | 5 | 4517 | ||
BC063307.1 | - | 5 | 4494 | 30340811 | ||
BM837454.1 | - | 1 | 482 | |||
BU933041.1 | - | 1 | 285 |