Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001430.4 | NP_001421.2 | 16 | 4910 | ||
XM_011532698.2 | XP_011531000.1 | 16 | 4668 | |||
GenBank mRNA | AF052094.1 | - | 1 | 993 | ||
AK123845.1 | - | 5 | 3373 | |||
BC015869.1 | - | 1 | 693 | |||
BC051338.1 | - | 16 | 4713 | 6305604 |