Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 12 | 2264 | ||
XM_011542647.2 | XP_011540949.1 | 11 | 1803 | |||
GenBank mRNA | AK057299.1 | - | 12 | 2264 | ||
AK223596.1 | - | 12 | 1653 | |||
AK296274.1 | - | 12 | 1543 | |||
AK297103.1 | - | 9 | 1010 | |||
AK299562.1 | - | 10 | 1101 | |||
AL832703.1 | - | 12 | 1635 | |||
BC039084.1 | - | 12 | 1633 | 4508489 | ||
X13822.1 | - | 6 | 597 | |||
Y00978.1 | - | 12 | 1509 |