Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002043.4 | NP_002034.3 | 9 | 1550 | ||
XM_011535713.2 | XP_011534015.1 | 8 | 2635 | |||
XM_011535714.2 | XP_011534016.1 | 8 | 2492 | |||
XM_011535715.2 | XP_011534017.1 | 8 | 2528 | |||
XM_011535716.2 | XP_011534018.1 | 8 | 3019 | |||
XM_011535717.2 | XP_011534019.1 | 8 | 2564 | |||
GenBank mRNA | AK293751.1 | - | 6 | 1248 | ||
BC130352.1 | - | 9 | 1469 | 40146383 | ||
BC130354.1 | - | 9 | 1469 | 40146385 | ||
DA863180.1 | - | 1 | 121 | |||
M86868.1 | - | 9 | 1496 |