Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002114.3 | NP_002105.3 | 4 | 705 | ||
XM_011514546.2 | XP_011512848.1 | 6 | 1748 | |||
XM_011514547.2 | XP_011512849.1 | 5 | 907 | |||
XM_011514548.2 | XP_011512850.1 | 5 | 1151 | |||
XM_011514549.2 | XP_011512851.1 | 5 | 1031 | |||
XM_011514550.2 | XP_011512852.1 | 5 | 898 | |||
XM_011514551.2 | XP_011512853.1 | 5 | 1576 | |||
XM_011514552.2 | XP_011512854.1 | 4 | 736 | |||
XM_011514553.1 | XP_011512855.1 | 4 | 717 | |||
XM_011514555.2 | XP_011512857.1 | 3 | 2306 | |||
XM_017010800.1 | XP_016866289.1 | 7 | 1634 | |||
XM_017010801.1 | XP_016866290.1 | 7 | 1575 | |||
XM_017010802.1 | XP_016866291.1 | 5 | 1654 | |||
XM_017010803.1 | XP_016866292.1 | 6 | 1749 | |||
XM_017010804.1 | XP_016866293.1 | 6 | 1752 | |||
GenBank mRNA | BC140816.1 | - | 3 | 638 | 9021684 | |
X51435.1 | - | 4 | 850 |