Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002185.3 | NP_002176.2 | 8 | 1544 | ||
NR_120485.1 | 6 | 1281 | ||||
XM_005248299.3 | XP_005248356.1 | 7 | 1435 | |||
GenBank mRNA | FJ529208.1 | - | 6 | 1267 | ||
M29696.1 | - | 8 | 1463 |