Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 24, 25 | 3079 | ||
NR_073103.1 | 23, 24 | 2976 | ||||
NR_073104.1 | 23, 24 | 2937 | ||||
NR_073105.1 | 24, 25 | 3120 | ||||
NR_073106.1 | 24, 25 | 3079 | ||||
NR_073107.1 | 23, 24 | 2958 | ||||
GenBank mRNA | BC143505.1 | - | 23, 24 | 2862 | ||
BC143508.1 | - | 23, 24 | 2823 | |||
BC143509.1 | - | 24, 25 | 3006 | |||
BC143511.1 | - | 24, 25 | 2965 | |||
BC143512.1 | - | 23, 24 | 2844 | |||
X17033.1 | - | 24, 25 | 2984 |